missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Biotin-VAD-FMK

Product Code. p-200104405
Encompass_Preferred
Click to view available options
Quantity:
1 mg
5 mg
Unit Size:
1mg
5mg
This item is not returnable. View return policy

Product Code. 16425947

Brand: MedChemExpress HY100894

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress Biotin-VAD-FMK is a cell permeable, irreversible biotin-labeled caspase inhibitor, used to identify active caspases in cell lysates.

Specifications

Quantity 5 mg
CAS 1135688-15-1
Product Type Biochemical Assay Reagents
Shipping Condition Room temperature
Solubility DMSO : ≥ 27.3 mg/mL (40.58 mM)
Storage Requirements Powder -20°C 3 years, 4°C 2 years ∣ In solvent -80°C 6 months, -20°C 1 month
Molecular Weight (g/mol) 672.81 g/mol
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.