missing translation for 'onlineSavingsMsg'
Learn More

MedChemExpress Biotin NHS

Product Code. 16431476
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 mg
200 mg
500 mg
Unit Size:
100mg
200mg
500mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16431476 200 mg 200mg
16428906 100 mg 100mg
16402326 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 16431476 Supplier MedChemExpress Supplier No. HYD0802
 Restricted Item
Estimated Shipment: 14-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress Biotin NHS is an amino reactive biotin reagent used in the preparation of biotinylated surfaces or polypeptides.

Specifications

Quantity 200 mg
Product Type Dye Reagents
Molecular Weight (g/mol) 341.38 g/mol
Solubility DMSO : 62.5 mg/mL (183.08 mM; Need ultrasonic)
Content And Storage -20°C, sealed storage, away from moisture and light ∣ The compound is unstable in solutions, freshly prepared is recommended.
Shipping Condition Room temperature
Target HBV
CAS 35013-72-0
Synonym Biotin-NHS ∣ Biotin N-hydroxysuccinimide ester ∣ NHS-Biotin
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.