missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Biotin-4-Fluorescein

Product Code. 11594137
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11594137 5 mg 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11594137 Supplier Invitrogen™ Supplier No. B10570
 Restricted Item
Estimated Shipment: 25-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Biotin-4-fluorescein can be used to quantitate available biotin-binding sites or the concentration of avidin or streptavidin through the strong quenching associated when this molecule binds to avidin.

Biotin-4-fluorescein can be used to quantitate available biotin-binding sites or the concentration of avidin or streptavidin through the strong quenching associated when this molecule binds to avidin.

To make a concentrated stock solution of up to 1 mM, dissolve in either anhydrous DMSO, DMF or 100% methanol. Stock solutions should be stored frozen, desiccated and protected from light.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Product Type Biotin-4-Fluorescein
Molecular Weight (g/mol) 644.7
Content And Storage Store at room temperature and protect from light.
Shipping Condition Room Temperature
Label or Dye FITC (Fluorescein)
Emission 523 nm
Excitation 494 nm

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.