Learn More
Agilent BioTek Microplate Washer Flat Magnets
For insertion into washer biomagnetic separation carriers
Brand: Agilent BioTek 7103016
Additional Details : Weight : 0.10000kg
Description
In flat-bottom well, beads pulled to band across well bottom. In round-bottom well, beads pulled to button at well bottom.
Order Info
For use with: 405 LS , 405 Touch, EL406, ELx405, ELx50 washers
Specifications
96-Well Flat Magnet | |
405 LS Microplate Washer, 405 Touch Microplate Washer, EL406 Microplate Washer, ELx 405 Microplate Washer, ELx 50 Microplate Strip Washer |