Learn More
Agilent BioTek Microplate Washer Flat Magnets
Shop All Agilent BioTek Products
Click to view available options
Type:
384-Well Flat Magnet
96-Well Flat Magnet
Unit Size:
Each
Description
In flat-bottom well, beads pulled to band across well bottom. In round-bottom well, beads pulled to button at well bottom.
Order Info
For use with: 405 LS , 405 Touch, EL406, ELx405, ELx50 washers
Specifications
Specifications
| Type | 384-Well Flat Magnet |
| For Use With (Equipment) | 405 LS Microplate Washer, 405 Touch Microplate Washer, EL406 Microplate Washer, ELx 405 Microplate Washer, ELx 50 Microplate Strip Washer |