Learn More
Agilent BioTek Complete Waste System
GREENER_CHOICE
Product Code. 15645437 Shop All Agilent BioTek Products
Click to view available options
Type:
Complete Dispense/High Flow Waste System
For Use With (Equipment):
EL406 Microplate Washer, ELx 405 Microplate Washer
ELx 405 Microplate Washer
Unit Size:
Each
Product Code. 15645437
Brand: Agilent BioTek 7100751
Provides supply and waste containment with optimal evacuation efficiency for 384- and 1536-well microplates
  • Includes complete high flow waste system and complete dispense system
  • 115V/230V
TRUSTED_SUSTAINABILITY
Specifications
Type Complete Dispense/High Flow Waste System
For Use With (Equipment) ELx 405 Microplate Washer