Learn More
Agilent BioTek Absorbance Test Plate
Product Code. 15810418 Shop All Agilent BioTek Products
Click to view available options
:
Absorbance test plate
Product Code. 15810418
Brand: Agilent BioTek 7260522
Includes
  • Exact spectrophotometric calibration data for each neutral density filter at eight wavelengths: 405, 450, 490, 550, 620, 630, 690, and 750nm
  • Protective case
Provides performance verification for microplate readers.
  • Tests alignment, accuracy, reproducibility and linearity
  • Conforms to the size and shape of a standard microplate to fit easily in a microplate reader
  • Ideal for monitoring an instrument's performance in compliance with regulatory requirements
Compliance
Uses NIST-traceable neutral density glass filters and precision machined blanking- and alignment-verification holes to confirm performance specifications.
TRUSTED_SUSTAINABILITY
Specifications
Wavelength 405; 450; 490; 550; 620; 630; 690; 750 nm