missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek BioStack Integration Kits
GREENER_CHOICE

Product Code. 10536234 Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
Biostack Microplate Stacker, 405 Touch Microplate Washer, 405 LS Microplate Washer
Biostack Microplate Stacker, EL406 Microplate Washer Dispenser
Biostack Microplate Stacker, ELx 405 Select Deep Well Microplate Washer
Unit Size:
Each
This item is not returnable. View return policy

Product Code. 10536234

Brand: Agilent BioTek 7310042

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Includes

Cat. No. BT7310042
Base for 90° and 0° integrations, four positioning feet, communication cable and interface software

Cat. No. BT7310041
Base for 90° and 0° integrations, four positioning feet and communication cable

Cat. No. BT7310040
Base for 90° and 0° integrations, four positioning feet and communication cable

Secures alignment for trouble-free performance.

  • Used with BioStack Microplate Stacker
  • Includes base for 90° and 0° integrations
  • Four positioning feet
TRUSTED_SUSTAINABILITY

Specifications

Includes Base for 90° and 0° integrations, 4 positioning feet and communication cable
Type Integration Kit
For Use With (Equipment) Biostack Microplate Stacker, EL406 Microplate Washer Dispenser
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.