missing translation for 'onlineSavingsMsg'
Learn More

Agilent BioTek BioStack Automated Microplate Stacker - Microplate Stacks
GREENER_CHOICE

Product Code. 15257639 Shop All Agilent BioTek Products
Click to view available options
Includes:
2 stacks of 10 microplates
2 stacks of 30 microplates
2 stacks of 50 microplates
Unit Size:
Each
Pack of 2
This item is not returnable. View return policy

Product Code. 15257639

Brand: Agilent BioTek 7310030

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Storage stacks

  • One set

TRUSTED_SUSTAINABILITY

Specifications

Product Type Microplates
For Use With (Equipment) Bio-tek Biostack automated microplate stacker, unlidded plates ≤14 mm
For Use With (Application) Storage
Includes 2 stacks of 10 microplates
Quantity 20 Microplates
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.