Learn More
Agilent BioTek BioSpa 8 Automated Incubator Accessory, Alignment kits
GREENER_CHOICE
Product Code. 16592132 Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
Epoch 2
Synergy H1 Microplate Readers
Unit Size:
Each
Product Code. 16592132
Brand: Agilent BioTek 1410009
For use with Imaging Multi-Mode Readers.
  • 11-120-552: for Synergy Neo2 and Cytation
  • 11-120-553: for Epoch 2
  • 11-120-554: for Synergy H1
TRUSTED_SUSTAINABILITY
Specifications
Product Type Alignment Kit
For Use With (Equipment) Epoch 2