Learn More
Agilent BioTek BioSpa 8 Automated Incubator Accessory, Alignment kits
For use with Imaging Multi-Mode Readers.
Brand:  Agilent BioTek 1410010
Additional Details : Weight : 1.70000kg
 View more versions of this product
Product Code. 16502142
  • £985.00 / Each
Estimated Shipment: 13-05-2024
to see stock.
Description
Description
  • 11-120-552: for Synergy Neo2 and Cytation
  • 11-120-553: for Epoch 2
  • 11-120-554: for Synergy H1
Specifications
Specifications
Alignment Kit
Synergy H1 Microplate Readers