Learn More
Sperian™ Honeywell™ Bionic Faceshield Visors
Product Code. 12655053
Change view
Click to view available options
Description:
Complete faceshield unit
Visor only
Lens Material:
Acetate
Polycarbonate
Lens Coating:
Anti-fog
Uncoated
Unit Size:
Each
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Description Lens Material Lens Coating Unit Size
12655053 Visor only Polycarbonate Anti-fog Each
12645053 Visor only Acetate Uncoated Each
12635083 Complete faceshield unit Acetate Uncoated Each
11826950 Complete faceshield unit Polycarbonate Uncoated Each
12635053 Visor only Polycarbonate - Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Product Code. 12655053 Supplier Sperian™ Supplier No. 1011627
Estimated Shipment: 10-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Offering maximum protection in an ergonomic design. Sperian™ Honeywell™ Bionic Faceshield Visors are incredibly adjustable, with 2784 possible wearer positions.
  • Extended top of head and chin protection
  • Large superior-quality visors for maximum visibility
  • Available with either polycarbonate visor for impact resistance or acetate visor for chemical protection
  • Comfortable fit with goggles or respirators
  • Locking mechanism for more complete safety
  • Dielectric design
  • Fully adjustable ratchet headgear and headband
  • Multiple tilt visor positions
  • Extra-soft cushioning on all parts of the shield in contact with the head
  • Breathable extra-soft sweatband is removable for easy washing and replacement
  • Excellent for the agriculture, chemical, forestry, steels and metals and wind energy industries
  • Protects against abrasion, airborne particles, biohazards, chemicals, impact, splashes and more
TRUSTED_SUSTAINABILITY
Specifications
Certifications/Compliance 89/686/EEC, EN166 / EN169 / EN170, ISO 9001 / 2000
Lens Coating Anti-fog
Description Visor only
Lens Tint Clear
Product Type Faceshield
Lens Material Polycarbonate