missing translation for 'onlineSavingsMsg'
Learn More

Beckman Coulter Life Sciences Biomek i-Series Pipette Tips

Product Code. 31903143 Shop All Beckman Coulter Life Sciences Products
Change view
Click to view available options
Volume (Metric):
50 μL
230 μL
1025 μL
Unit Size:
Pack of 480
Pack of 960
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
31903143 1025 μL Pack of 480
31903145 50 μL Pack of 960
31903144 230 μL Pack of 960
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 31903143 Supplier Beckman Coulter Life Sciences Supplier No. B85955
Estimated Shipment: 17-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For use with Biomek liquid handlers and contamination-free liquid handling performance for automated workflows.

  • Designed, validated, and approved specifically for Beckman Coulter Biomek i-Series liquid handling systems for optimal compatibility and performance
  • Available in multiple volume capacities and configurations including conductive, filtered, sterile, and wide-bore options to support diverse laboratory applications
  • Manufactured from premium-grade virgin polypropylene
  • Color-coded racks with integrated tip definitions and pipetting techniques in Biomek software help streamline method development and workflow setup
  • Sterile options are processed using validated sterilization methods for sensitive genomics, proteomics, and molecular biology applications

Specifications

Volume (Metric) 1025 μL
Filtered Filtered
Sterility Sterile
Color Clear
Autoclavable Not Autoclavable
Format Box
Tip Style Finely Tapered, Barrier
Quantity 480 Tips
Product Type Tip Box
For Use With (Equipment) Beckman Coulter Liquid Handling Systems
Material 100% Virgin Polypropylene
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.