missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Biocytin (ε-Biotinoyl-L-Lysine)

Product Code. 11594157
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
11594157 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 
Product Code. 11594157 Supplier Invitrogen™ Supplier No. B1592
 Restricted Item
Estimated Shipment: 09-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
For Research Use Only. All usage must comply with product instructions.
 

Biocytin (ε-Biotinoyl-L-Lysine)

Biocytin can be used as microinjectable anterograde and transneuronal tracers that is fixable with aldehyde-based fixatives. Additionally the primary amine on the lysine can be reversibly coupled to aldehydes and ketones to form a Schiff base - which can be reduced to a generate stable amine derivative by sodium borohydride (NaBH4) or sodium cyanoborohydride (NaCNH3). Carboxylic acids of proteins and other water-soluble biopolymers can be coupled to this molecule in aqueous solution using water-soluble carbodiimides such as EDAC (E2247).
TRUSTED_SUSTAINABILITY

Specifications

Product Type Biocytin
Quantity 100 mg
Content And Storage Store at room temperature.
Reagent Type Biocytins
Shipping Condition Room Temperature
Label Type Biotin & Other Haptens

For Research Use Only. Not for use in diagnostic procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.