missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Poly-D-Lysine Vented Cap Flasks
GREENER_CHOICEBIOPHARMA

Product Code. 10666111
Change view
Click to view available options
Culture Area:
150 cm2
175 cm2
25 cm2
75 cm2
No. per Case:
10/cs.
40/cs.
5/cs.
50/cs.
Unit Size:
Pack of 10
Pack of 40
Pack of 5
Pack of 50
8 product options available for selection
Product selection table with 8 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Culture Area No. per Case unitSize
10666111 75 cm2 5/cs. Pack of 5
10297280 150 cm2 5/cs. Pack of 5
10626691 175 cm2 5/cs. Pack of 5
10796672 75 cm2 50/cs. Pack of 50
10410102 175 cm2 40/cs. Pack of 40
10397941 25 cm2 50/cs. Pack of 50
10400102 150 cm2 40/cs. Pack of 40
10711371 25 cm2 10/cs. Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 8 options available.
8 options
This item is not returnable. View return policy
Product Code. 10666111 Supplier Corning™ Supplier No. 354537

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines

Corning™ BioCoat™ products come in a variety of pre-coated options and are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures.
TRUSTED_SUSTAINABILITY

Specifications

Capacity (English) 8.45 oz.
Closure Material Phenolic
Capacity (Metric) 250 mL
For Use With (Application) For adhesion, growth and differentiation of neuronal and transfected cell lines
Neck Style Canted
No. per Case 5/cs.
Shape Rectangular
Surface Coating Poly-D-Lysine
Sterility Sterile
Culture Area 75 cm2

Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.