missing translation for 'onlineSavingsMsg'
Learn More

Corning™ BioCoat™ Poly-D-Lysine Vented Cap Flasks

Ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines

Brand:  Corning™ 354538

Code : 67

Additional Details : Weight : 0.87000kg

 View more versions of this product

Product Code. 10297280

  • £104.62 / Pack of 5
Estimated Shipment: 27-08-2024
to see stock.
Add to basket

Description

Description

Corning™ BioCoat™ products come in a variety of pre-coated options and are an ideal solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures.
Specifications

Specifications

20.2 oz.
600 mL
Canted
Rectangular
Sterile
Phenolic
For adhesion, growth and differentiation of neuronal and transfected cell lines
5/cs.
Poly-D-Lysine
150 cm2
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Disclaimer: This product is not intended to mitigate the presence of microorganisms on surfaces or in the environment, where such organisms can be deleterious to humans or the environment.