missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Corning™ BioCoat™ Collagen IV Rectangular Canted Neck Culture Flask
Designed to closely mimic in vivo environments and enhance expression of cell-specific morphology. Corning™ BioCoat™ Collagen IV Rectangular Canted Neck Culture Flask is ideal for applications involving adhesion, growth and differentiation of neuronal and transfected cell lines.
£277.11 - £430.86
Specifications
Closure Material | Phenolic |
---|---|
For Use With (Application) | For adhesion, growth and differentiation of neuronal and transfected cell lines |
Neck Style | Canted |
Shape | Rectangular |
Surface Coating | Collagen IV |
Product Code | Brand | Capacity (Metric) | Culture Area | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Capacity (Metric) | Culture Area | Price | Quantity & Availability | ||||
10070741
|
Corning™
354523 |
250 mL | 75 cm2 |
£419.00
Pack of 10 |
In Stock
Log in to see stock. |
||||
10346741
|
Corning™
354528 |
750 mL | 175 cm2 |
£430.86
Pack of 5 |
Estimated Shipment: 12-09-2024
Log in to see stock. |
||||
10102711
|
Corning™
354534 |
70 mL | 25 cm2 |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
|||||
Description
- BioCoat™ provides a superior solution for enhanced cell attachment and growth of a variety of primary cells and transformed cells in serum-free or serum-containing cultures
- Matrix components are applied as an even, optically clear coating
- All lots tested and found negative for bacteria and fungi; cellware is tested for the ability to promote firm attachment of rat cerebellar granule cells
For research use only.
Specifications
Phenolic | |
Canted | |
Collagen IV | |
Plug Seal |
For adhesion, growth and differentiation of neuronal and transfected cell lines | |
Rectangular | |
Sterile |