Learn More
Ansell BioClean-D™ Non-sterile Cleanroom Coverall with Hood and Integrated Boots
Product Code. 30058505
Change view
Click to view available options
Size:
X-Small
Unit Size:
Pack of 20
Product Code. Size Unit Size
30058505 X-Small Pack of 20
1 options
Product Code. 30058505 Supplier Ansell Supplier No. BDFCXS
  • ONLINE EXCLUSIVE PRICE
    £274.10
    £153.18 / Pack of 20
    Save £120.92
    44% Off
Estimated Shipment: 04-12-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Cleanroom Coverall made of CleanTough™ fabric for lightweight, low-linting body protection with front zip and protective flap to provide contaminant and liquid splash barrier.
  • Reduced contamination risks: The BioClean-D Coverall with Hood and Integrated Boots BDFC is low-linting and ISO cleanroom-compliant thanks to its lightweight CleanTough™ composition, ensuring contamination risks are kept to a minimum
  • Optimized protection: This non-sterile disposable coverall features a front zip with a protective flap, which guards against liquid splashes and chemicals, while thumbs loops and an elasticated hood, back, cuffs and ankles offer a secure fit
  • Enhanced features: This antistatic coverall is also equipped with integrated boots and slip-resistant soles, for added workplace safety in laboratory environments
  • Reduced contamination risks: The BioClean-D Coverall with Hood and Integrated Boots BDFC is low-linting and ISO cleanroom-compliant thanks to its lightweight CleanTough™ composition, ensuring contamination risks are kept to a minimum
  • Optimized protection: This non-sterile disposable coverall features a front zip with a protective flap, which guards against liquid splashes and chemicals, while thumbs loops and an elasticated hood, back, cuffs and ankles offer a secure fit
  • Enhanced features: This antistatic coverall is also equipped with integrated boots and slip-resistant soles, for added workplace safety in laboratory environments
TRUSTED_SUSTAINABILITY
Specifications
Product Type Cleanroom Coveralls
Size X-Small
Gender Unisex
Material Flashspun Polyethylene
Color White
Disposable Single-use
Waist Style Non-elastic
Ankle Style Attached Boots
Cuff Style Elastic
Neck Style Hooded
Hood Type Attached, Elastic Face
Closure Type Front Zipper with Storm Flap
Seam Type Bound
Sterility Non-sterile
Resistance Anti-static, Chemicals, Liquid, Particle
Collar Type Hooded
Certifications/Compliance (Europe) ASTM F739, CE 0598, EN ISO 13688:2013, EN 1149-5:2008, EN 13934-1, EN 13935-2, EN 530, EN 6530, EN 7854, EN 863, EN 9073-4, EN ISO 14325, Category III, EN ISO 13982-1:2004 + A1:2010, EN 13034:2005 + A1:2009
Latex Free No
No. of Pockets None
For Use With (Application) Blending, compounding solids or liquids, Crime Scene Investigation (CSI), Forensics, Handling of Powders, Aseptic Filling and Labelling, Cleanroom Cleaning and Preparing, Medical Device Manufacturing, Biotechnologies Blending Operations, Handling Active Material, Maintaining Equipment and Instruments
Cleanroom Class Class 10 (ISO 4)