missing translation for 'onlineSavingsMsg'
Learn More

Ansell BioClean™ BAFS 601 Anti-Fog Goggle Spray

Product Code. 30245467
Change view
Click to view available options
Quantity:
60 mL
Unit Size:
Pack of 10
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30245467 60 mL Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
N/A
Product Code. 30245467 Supplier Ansell Supplier No. BAFS601
 Restricted Item
In Stock
Add to basket
Edge
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
N/A

Alcohol-free with airless pump action spray for use with autoclavable goggles.

  • Anti-fog effect: For less obstruction when using autoclavable goggles
  • Airless pump action: Quick and easy application
  • Manufactured at an ISO 9001-certified facility: For assured quality
TRUSTED_SUSTAINABILITY

Specifications

Lens Coating Anti-fog
Color Clear
Product Type Lens Cleaning Solution
For Use With (Application) Address fogging issues when using autoclavable goggles
No. per Case 100/Cs.
Model BAFS
Quantity 60 mL
No. per Pack 10/Pk.
Size Universal
Material Proprietary Formula
Disposable Single-use
Autoclavable Not Autoclavable
Show More Show Less

N/A

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.