missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BioBasic™ 8 HPLC Columns
Use this column for reversed-phase separation of proteins and other biomolecules.
£240.00 - £727.62
Specifications
Max. Pressure | 5800 psi (400 bar) |
---|---|
Particle Size | 5 μm |
pH | 2 to 9 |
Pore Size | 300 Å |
Surface Area | 100 m²/g |
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
10756173
|
Thermo Scientific™
72205-254630 |
5 μm | 4.6 mm | 250 mm |
£727.62
Each |
Estimated Shipment: 13-05-2024
Log in to see stock. |
|||
10161213
|
Thermo Scientific™
72205-052130 |
5 μm | 2.1 mm | 50 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10445553
|
Thermo Scientific™
72205-101030 |
5 μm | 1 mm | 100 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10434393
|
Thermo Scientific™
72205-104630 |
5 μm | 4.6 mm | 100 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10362595
|
Thermo Scientific™
72205-151030 |
5 μm | 1 mm | 150 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10293353
|
Thermo Scientific™
72205-152130 |
5 μm | 2.1 mm | 150 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10253313
|
Thermo Scientific™
72205-154630 |
5 μm | 4.6 mm | 150 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10283273
|
Thermo Scientific™
72205-051030 |
5 μm | 1 mm | 50 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10424393
|
Thermo Scientific™
72205-102130 |
5 μm | 2.1 mm | 100 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11558320
|
Thermo Scientific™
72205-014001 |
5 μm | 4 mm | 10 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
11548320
|
Thermo Scientific™
72205-012101 |
5 μm | 2.1 mm | 10 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10311505
|
Thermo Scientific™
72205-011001 |
5 μm | 1 mm | 10 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Optimized for the separation of a wide range of peptides, Thermo Scientific™ BioBasic™ 8 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules.
- 300Å pore size for improved biomolecule analysis
- An excellent starting column for protein and peptide analysis
- Outstanding reproducibility, efficiency and column lifetime
- Excellent for LC/MS separations
Ordering Information:
Other column dimensions are available, including preparative columns. Please call Customer Service for more information.
Specifications
5800 psi (400 bar) | |
2 to 9 | |
100 m²/g | |
L7 | |
5% | |
Yes |
5 μm | |
300 Å | |
60°C | |
BioBasic | |
Reversed Phase | |
Reversed Phase |