Learn More
Thermo Scientific™ BioBasic™ 8 HPLC Columns
Use this column for reversed-phase separation of proteins and other biomolecules.
Brand: Thermo Scientific™ 72205-102130
Code : 16
Additional Details : Weight : 0.10000kg
Description
Optimized for the separation of a wide range of peptides, Thermo Scientific™ BioBasic™ 8 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules.
- 300Å pore size for improved biomolecule analysis
- An excellent starting column for protein and peptide analysis
- Outstanding reproducibility, efficiency and column lifetime
- Excellent for LC/MS separations
Ordering Information:
Other column dimensions are available, including preparative columns. Please call Customer Service for more information.
Specifications
Analytical Column | |
100 mm | |
5 μm | |
300 Å | |
60°C | |
C8 | |
Spherical, Fully Porous, Ultrapure Silica | |
Reversed Phase | |
Reversed Phase |
2.1 mm | |
5800 psi (400 bar) | |
2 to 9 | |
100 m²/g | |
L7 | |
BioBasic | |
5% | |
Yes | |
Analytical Column |