Learn More
Thermo Scientific™ BioBasic™ SEC 60 Size Exclusion Chromatography HPLC Columns
Outstanding size separation of water soluble compounds.
Brand: Thermo Scientific™ 73305-307846
Code : 16
Additional Details : Weight : 0.30000kg
Description
Provide outstanding size separation of water soluble compounds with Thermo Scientific™ BioBasic™ SEC 60 Size Exclusion Chromatography HPLC Columns. BioBasic SEC columns provide high efficiency separations for a wide range of molecular weights from 100 to 10,000,000 daltons, allowing the analysis of proteins, peptides and other biomolecules.
- Covers separation of analytes over a wide molecular weight range
- Long column life and high column efficiencies
- Simple mechanism of interaction based on molecular size and shape
- Excellent for sample cleanup
- Straightforward method development, simple mobile phases
- 60Å pore size
- Particle size: 5μm
Ordering Information:
For information on bulk quantities, please inquire.
Specifications
High Temperature Column | |
Size Exclusion Chromatography | |
400 bar | |
60 Å | |
Diol | |
Silica, Spherical Fully Porous Ultrapure | |
High Temperature Column |
7.8 mm | |
300 mm | |
5 μm | |
L33 | |
BioBasic | |
Size Exclusion |