missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific™ BioBasic™ SEC 60 Size Exclusion Chromatography HPLC Columns
Outstanding size separation of water soluble compounds.
£476.00 - £1320.00
Specifications
Diameter (Metric) | 7.8 mm |
---|---|
For Use With | Size Exclusion Chromatography |
Particle Size | 5 μm |
Pore Size | 60 Å |
USP Type | L33 |
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
10785526
|
Thermo Scientific™
73305-157846 |
5 μm | 7.8 mm | 150 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10354613
|
Thermo Scientific™
73305-307846 |
5 μm | 7.8 mm | 300 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
10669295
|
Thermo Scientific™
73305-037821 |
5 μm | 7.8 mm | 30 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Provide outstanding size separation of water soluble compounds with Thermo Scientific™ BioBasic™ SEC 60 Size Exclusion Chromatography HPLC Columns. BioBasic SEC columns provide high efficiency separations for a wide range of molecular weights from 100 to 10,000,000 daltons, allowing the analysis of proteins, peptides and other biomolecules.
- Covers separation of analytes over a wide molecular weight range
- Long column life and high column efficiencies
- Simple mechanism of interaction based on molecular size and shape
- Excellent for sample cleanup
- Straightforward method development, simple mobile phases
- 60Å pore size
- Particle size: 5μm
Ordering Information:
For information on bulk quantities, please inquire.
Specifications
7.8 mm | |
5 μm | |
L33 | |
Size Exclusion |
Size Exclusion Chromatography | |
60 Å | |
BioBasic |