Learn More
Thermo Scientific™ BioBasic™ 4 HPLC Columns
Use this 300Å wide pore silica for analysis of larger hydrophobic proteins and peptides that bind too tightly on C8 or C18 columns.
£363.32 - £769.50
Specifications
Max. Pressure | 5800 psi (400 bar) |
---|---|
Particle Size | 5 μm |
pH | 2 to 9 |
Pore Size | 300 Å |
Surface Area | 100 m²/g |
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Particle Size | Diameter (Metric) | Length (Metric) | Price | Quantity & Availability | |||
10583995
|
Thermo Scientific™
72305-051030 |
5 μm | 1 mm | 50 mm |
£769.50
Each |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||
10263313
|
Thermo Scientific™
72305-052130 |
5 μm | 2.1 mm | 50 mm |
£697.58
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||
10395003
|
Thermo Scientific™
72305-101030 |
5 μm | 1 mm | 100 mm |
£754.72
Each |
Estimated Shipment: 16-05-2024
Log in to see stock. |
|||
10581105
|
Thermo Scientific™
72305-102130 |
5 μm | 2.1 mm | 100 mm |
£688.48
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||
10692854
|
Thermo Scientific™
72305-104630 |
5 μm | 4.6 mm | 100 mm |
£725.15
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||
10110693
|
Thermo Scientific™
72305-152130 |
5 μm | 2.1 mm | 150 mm |
£724.62
Each |
Estimated Shipment: 17-05-2024
Log in to see stock. |
|||
10384513
|
Thermo Scientific™
72305-154630 |
5 μm | 4.6 mm | 150 mm |
£690.49
Each |
Estimated Shipment: 12-06-2024
Log in to see stock. |
|||
10344563
|
Thermo Scientific™
72305-252130 |
5 μm | 2.1 mm | 250 mm |
£763.75
Each |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||
10541305
|
Thermo Scientific™
72305-254630 |
5 μm | 4.6 mm | 250 mm |
£727.62
Each |
In Stock
Log in to see stock. |
|||
11598320
|
Thermo Scientific™
72305-012101 |
5 μm | 2.1 mm | 10 mm |
£402.86
Pack of 4 |
Estimated Shipment: 15-05-2024
Log in to see stock. |
|||
11508330
|
Thermo Scientific™
72305-014001 |
5 μm | 4 mm | 10 mm |
£363.32
Pack of 4 |
In Stock
Log in to see stock. |
|||
10649585
|
Thermo Scientific™
72305-151030 |
5 μm | 1 mm | 150 mm |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||
Description
Designed for the analysis of larger proteins and peptides, Thermo Scientific™ BioBasic™ 4 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules with high hydrophobicity.
- Based on 300Å silica for outstanding biomolecule performance
- Lower carbon load for optimal retention of larger peptides and proteins
- Outstanding reproducibility, efficiency and column lifetime
- Excellent for LC/MS separations
Ordering Information:
Other column dimensions are available, including preparative columns. Please call your Customer Service Representative for more information.
Specifications
5800 psi (400 bar) | |
2 to 9 | |
100 m²/g | |
L26 | |
4% | |
Yes |
5 μm | |
300 Å | |
60°C | |
C4 | |
Reversed Phase | |
Reversed Phase |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.