Learn More
Thermo Scientific™ BioBasic™ 4 HPLC Columns
Use this 300Å wide pore silica for analysis of larger hydrophobic proteins and peptides that bind too tightly on C8 or C18 columns.
Brand: Thermo Scientific™ 72305-104630
Code : 16
Additional Details : Weight : 0.10000kg
Description
Designed for the analysis of larger proteins and peptides, Thermo Scientific™ BioBasic™ 4 LC Columns are high-performance columns ideal for the analysis of proteins, peptides and other biomolecules with high hydrophobicity.
- Based on 300Å silica for outstanding biomolecule performance
- Lower carbon load for optimal retention of larger peptides and proteins
- Outstanding reproducibility, efficiency and column lifetime
- Excellent for LC/MS separations
Ordering Information:
Other column dimensions are available, including preparative columns. Please call your Customer Service Representative for more information.
Specifications
Analytical Column | |
100 mm | |
5 μm | |
300 Å | |
60°C | |
C4 | |
Spherical, Fully Porous, Ultrapure Silica | |
Reversed Phase | |
Reversed Phase | |
Bio-LC |
4.6 mm | |
5800 psi (400 bar) | |
2 to 9 | |
100 m²/g | |
L26 | |
BioBasic | |
4% | |
Yes | |
Analytical Column |
Limited warranty period: Subject to all warranty requirements and exclusions set out in our Terms and Conditions of Sale, this product is warranted from the date we ship the product and for ninety (90) days thereafter. Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.