missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Tocris Bioscience™ BIO

Click to view available options
Quantity:
10 mg
50 mg
Unit Size:
10mg
50mg
Description
Potent ATP-competitive glycogen synthase kinase-3 (GSK-3) inhibitor (IC50 = 5 nM). Also inhibits cyclin-dependent kinases (cdks; IC50 values are <100 nM for cdk2 and 5 and sub micromolar for cdk1, cdk4 and cdk9). Maintains self-renewal and pluripotency in embryonic stem cells via activation of the Wnt signaling pathway in vitro; also promotes proliferation and dedifferentiation in cardiomyocytes.
Negative Control also available.
Specifications
Specifications
| Quantity | 10 mg |
| Formula Weight | 356.17 |
| Purity | >98% |
| Molecular Weight (g/mol) | 356.179 |
| Chemical Name or Material | BIO |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction