CAS RN 667463-62-9
CAS RN 667463-62-9
IUPAC Name:
6'-bromo-3-(hydroxyamino)-1',2'-dihydro-[2,3'-biindolyliden]-2'-one
Synonyms:
6'-bromo-3-(hydroxyamino)-1'H-[2,3'-biindolyliden]-2'-one
Molecular Weight (g/mol):
356.18
Molecular Formula:
C16H10BrN3O2
InChi Key:
WNWSUJQVZJJGLF-UHFFFAOYSA-N
SMILES:
ONC1=C2C=CC=CC2=NC1=C1C(=O)NC2=C1C=CC(Br)=C2
1
–
3
of
3
results
1
Tocris Bioscience™ BIO
CAS: 667463-62-9 Molecular Formula: C16H10BrN3O2
2
3
GSK 3 Inhibitor IX, MedChemExpress
MedChemExpress GSK 3 Inhibitor IX (6-Bromoindirubin-3'-oxime; BIO) is a potent, selective, reversible and ATP-competitive inhibitor of GSK-3α/β and CDK1-cyclinB complex with IC50s of 5 nM/320 nM/80 nM for (GSK-3α/β)/CDK1/CDK5, respectively.