Learn More
Hirschmann™ BIO-SILICO Silicone Stoppers
Excellent for the cultivation of aerobic microorganisms. Hirschmann™ SILICOSEN Silicone Stoppers are specially processed culture plugs with coarse bubbles for preparation, filling and sterilization of culture media.
Brand: Hirschmann™ 8905917
Code : A0
Additional Details : Weight : 0.80000kg
Description
- Coarse pore structure
- Good air permeability
- Chemically and temperature resistant
- Liquid repellent and reusable
Specifications
Orange | |
13-16 mm | |
10/Pk. | |
15 mm |
Stopper | |
For The Preparation And Sterilization Of Culture Media | |
Silicone |