missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sartorius™ BF2 Cotton Linter Blotting Paper
Highly pure and ideal for blotting and chromatography, as well as a wide range of absorption applications like those common in the life sciences and diagnostics sectors.
£104.00 - £365.00
Specifications
Color | White |
---|---|
Quantity | 100/Pk. |
Material | Cotton linters with an alpha-cellulose content |
Thickness | 0.35 mm |
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Length (Metric) | Price | Quantity & Availability | |||||
10525891
|
Sartorius™
FT-2-519-200200N |
200 mm |
£104.00
Pack of 100 |
Estimated Shipment: 24-06-2024
Log in to see stock. |
|||||
10317691
|
Sartorius™
FT-2-519-460570N |
460 mm |
£321.00
Pack of 100 |
Estimated Shipment: 13-06-2024
Log in to see stock. |
|||||
10490652
|
Sartorius™
FT-2-519-580600N |
580 mm |
£365.00
Pack of 100 |
In Stock
Log in to see stock. |
|||||
Description
- Grade BF 2
- Made of high purity cotton linters for uniform buffer flow and resulting blots
- No additives to avoid any interference during the transfer
- Supplied in sheets
Specifications
White | |
100/Pk. | |
Cotton linters with an alpha-cellulose content | |
0.35 mm |