missing translation for 'onlineSavingsMsg'
Learn More

Sartorius™ BF2 Cotton Linter Blotting Paper

Product Code. 10490652 Shop All Sartorius Products
Click to view available options
Length (Metric):
200 mm
460 mm
580 mm
Unit Size:
Pack of 100
This item is not returnable. View return policy

Product Code. 10490652

Brand: Sartorius™ FT2519580600N

Only null left
Add to basket
Add to basket
This item is not returnable. View return policy

Highly pure and ideal for blotting and chromatography, as well as a wide range of absorption applications like those common in the life sciences and diagnostics sectors.

  • Grade BF 2
  • Made of high purity cotton linters for uniform buffer flow and resulting blots
  • No additives to avoid any interference during the transfer
  • Supplied in sheets
TRUSTED_SUSTAINABILITY

Specifications

Length (Metric) 580 mm
Width (Metric) 600 mm
Color White
Quantity 100/Pk.
Material Cotton linters with an alpha-cellulose content
Thickness 0.35 mm
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.