missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta Tubulin Antibody (4E4), Novus Biologicals™
Mouse Monoclonal Antibody
£175.00 - £388.00
Specifications
| Antigen | beta Tubulin |
|---|---|
| Clone | 4E4 |
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:5000 - 1:10000, Immunohistochemistry 1:1000-1:5000, Immunocytochemistry/ Immunofluorescence 1:1000 - 1:5000 |
| Applications | Western Blot, Immunohistochemistry, Immunofluorescence |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18654467
|
Novus Biologicals
NBP2-50047 |
0.1 mL |
£388.00
0.10mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18645457
|
Novus Biologicals
NBP2-50047SS |
0.025 mL |
£175.00
0.03mL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
beta Tubulin Monoclonal antibody specifically detects beta Tubulin in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ ImmunofluorescenceSpecifications
| beta Tubulin | |
| 1 mg/mL | |
| Western Blot, Immunohistochemistry, Immunofluorescence | |
| Unconjugated | |
| Mouse | |
| Apoptosis, Breast Cancer, Cell Biology, Cellular Markers, Signal Transduction | |
| 50% PBS, 50% glycerol | |
| beta polypeptide, MGC117247, Tubb5, tubulin beta chain, tubulin beta polypeptide, tubulin beta-1 chain, Tubulin beta-5 chain, tubulin, beta | |
| Pig brain beta Tubulin. [UniProt# Q767L7] | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
| 4E4 | |
| Western Blot 1:5000 - 1:10000, Immunohistochemistry 1:1000-1:5000, Immunocytochemistry/ Immunofluorescence 1:1000 - 1:5000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| Human, Mouse, Rat, Monkey | |
| P07437 | |
| 203068 | |
| IgG2a | |
| Immunogen affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title