missing translation for 'onlineSavingsMsg'
Learn More

beta Tubulin Antibody (4E4), Novus Biologicals™

Product Code. 18654467 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.025 mL
0.1 mL
Unit Size:
0.03mL
0.1mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18654467 0.1 mL 0.1mL
18645457 0.025 mL 0.03mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 18654467 Supplier Novus Biologicals Supplier No. NBP250047

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

beta Tubulin Monoclonal antibody specifically detects beta Tubulin in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY

Specifications

Antigen beta Tubulin
Applications Western Blot, Immunohistochemistry, Immunofluorescence
Classification Monoclonal
Clone 4E4
Concentration 1 mg/mL
Conjugate Unconjugated
Dilution Western Blot 1:5000 - 1:10000, Immunohistochemistry 1:1000-1:5000, Immunocytochemistry/ Immunofluorescence 1:1000 - 1:5000
Formulation 50% PBS, 50% glycerol
Gene Accession No. P07437
Gene Alias beta polypeptide, MGC117247, Tubb5, tubulin beta chain, tubulin beta polypeptide, tubulin beta-1 chain, Tubulin beta-5 chain, tubulin, beta
Host Species Mouse
Immunogen Pig brain beta Tubulin. [UniProt# Q767L7]
Purification Method Immunogen affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Apoptosis, Breast Cancer, Cell Biology, Cellular Markers, Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 203068
Target Species Human, Mouse, Rat, Monkey
Content And Storage Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
Form Purified
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.