missing translation for 'onlineSavingsMsg'
Learn More

beta Tubulin Antibody (1B12), Novus Biologicals™

Mouse Monoclonal Antibody

£186.00 - £407.00

Specifications

Antigen beta Tubulin
Clone 1B12
Concentration 1 mg/mL
Dilution Western Blot 1:5000 - 1:10000, Immunocytochemistry/ Immunofluorescence 1:5000, Immunofluorescence, Single Cell Western
Applications Western Blot, Immunofluorescence, Immunofluorescence, Single Cell Western
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18668806
View Documents
Novus Biologicals
NBP2-50061SS
0.025 mL
£186.00
0.03mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18637898
View Documents
Novus Biologicals
NBP2-50061
0.1 mL
£407.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

beta Tubulin Monoclonal antibody specifically detects beta Tubulin in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunofluorescence, Single Cell Western
Specifications

Specifications

beta Tubulin
1 mg/mL
Western Blot, Immunofluorescence, Immunofluorescence, Single Cell Western
Unconjugated
Mouse
Apoptosis, Breast Cancer, Cell Biology, Cellular Markers, Signal Transduction
50% PBS, 50% glycerol
beta polypeptide, MGC117247, Tubb5, tubulin beta chain, tubulin beta polypeptide, tubulin beta-1 chain, Tubulin beta-5 chain, tubulin, beta
Pig brain beta Tubulin preparation. [UniProt# Q767L7]
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles.
1B12
Western Blot 1:5000 - 1:10000, Immunocytochemistry/ Immunofluorescence 1:5000, Immunofluorescence, Single Cell Western
Monoclonal
Purified
RUO
Human, Mouse, Rat, Monkey
P07437
203068
IgG2b
Immunogen affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.