missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
beta Tubulin Monoclonal antibody specifically detects beta Tubulin in Human, Mouse, Rat, Monkey samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence, Immunofluorescence, Single Cell Western
Specifications
Specifications
| Antigen | beta Tubulin |
| Applications | Western Blot, Immunofluorescence, Immunofluorescence, Single Cell Western |
| Classification | Monoclonal |
| Clone | 1B12 |
| Concentration | 1 mg/mL |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:5000 - 1:10000, Immunocytochemistry/ Immunofluorescence 1:5000, Immunofluorescence, Single Cell Western |
| Formulation | 50% PBS, 50% glycerol |
| Gene Accession No. | P07437 |
| Gene Alias | beta polypeptide, MGC117247, Tubb5, tubulin beta chain, tubulin beta polypeptide, tubulin beta-1 chain, Tubulin beta-5 chain, tubulin, beta |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?