missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific Chemicals beta-Lapachone, 98%

Product Code. 16656032
Change view
Click to view available options
Quantity:
5 mg
Unit Size:
5mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16656032 5 mg 5mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16656032 Supplier Thermo Scientific Chemicals Supplier No. J67308.#0
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

 

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Specifications

Quantity 5 mg
Description Orange
Content And Storage -30°C to -10°C
CAS 4707-32-8
Molecular Formula C15H14O3
Formula Weight 242.27
Melting Point 151°C to 153°C
Product Type beta-Lapachone, 98%
Form Solid
Assay 98%

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.