missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Beta Hydroxysteroid Dehydrogenase Antibody - Azide and BSA Free, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-92640-0.1ml
This item is not returnable.
View return policy
Description
Beta Hydroxysteroid Dehydrogenase Polyclonal antibody specifically detects Beta Hydroxysteroid Dehydrogenase in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications
| Beta Hydroxysteroid Dehydrogenase | |
| Polyclonal | |
| Western Blot 1:100 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10-1:100 | |
| 3 beta-HSD type II, 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 2, 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type II, 3-beta-HSD II, delta 5-delta 4-isomerase type II, HSD3B, HSDB, HSDB3B, hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 2,3-beta-hydroxy-Delta(5)-steroid dehydrogenase, progesterone reductase, SDR11E2, short chain dehydrogenase/reductase family 11E, member 2,3-beta-hydroxy-5-ene steroid dehydrogenase | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 1-287 of human HSD3B2 (NP_000189.1). MGWSCLVTGAGGLLGQRIVRLLVEEKELKEIRALDKAFRPELREEFSKLQNRTKLTVLEGDILDEPFLKRACQDVSVVIHTACIIDVFGVTHRESIMNVNVKGTQLLLEACVQASVPVFIYTSSIEVAGPNSYKEIIQNGHEEEPLENTWPTPYPYSKKLAEKAVLAANGWNLKNGDTLYTCALRPTYIYGEGGPFLSASINEALNNNGILSSVGKFSTVNPVYVGNVAWAHILALRALRDPKKAPSVRGQFYYISDDTPHQSYDNLNYILSKEFGLRLDSRWSLPL | |
| 0.1 mL | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, Immunofluorescence | |
| Unconjugated | |
| PBS with 50% glycerol, pH7.3. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 3284 | |
| Store at -20°C. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction