missing translation for 'onlineSavingsMsg'
Learn More

Beta Hydroxysteroid Dehydrogenase Antibody - Azide and BSA Free, Novus Biologicals™

Rabbit Polyclonal Antibody

£151.00 - £359.00

Specifications

Antigen Beta Hydroxysteroid Dehydrogenase
Dilution Western Blot 1:100 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10-1:100
Applications Western Blot, Immunofluorescence
Classification Polyclonal
Conjugate Unconjugated
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18648312
View Documents
Novus Biologicals
NBP2-92640-0.02ml
0.02 mL
£151.00
0.02mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
18684962
View Documents
Novus Biologicals
NBP2-92640-0.1ml
0.1 mL
£359.00
0.10mL
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Beta Hydroxysteroid Dehydrogenase Polyclonal antibody specifically detects Beta Hydroxysteroid Dehydrogenase in Human, Mouse samples. It is validated for Western Blot, Immunocytochemistry/ Immunofluorescence
Specifications

Specifications

Beta Hydroxysteroid Dehydrogenase
Western Blot, Immunofluorescence
Unconjugated
Rabbit
Human, Mouse
3 beta-HSD type II, 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type 2, 3 beta-hydroxysteroid dehydrogenase/Delta 5-->4-isomerase type II, 3-beta-HSD II, delta 5-delta 4-isomerase type II, HSD3B, HSDB, HSDB3B, hydroxy-delta-5-steroid dehydrogenase, 3 beta- and steroid delta-isomerase 2,3-beta-hydroxy-Delta(5)-steroid dehydrogenase, progesterone reductase, SDR11E2, short chain dehydrogenase/reductase family 11E, member 2,3-beta-hydroxy-5-ene steroid dehydrogenase
Recombinant fusion protein containing a sequence corresponding to amino acids 1-287 of human HSD3B2 (NP_000189.1). MGWSCLVTGAGGLLGQRIVRLLVEEKELKEIRALDKAFRPELREEFSKLQNRTKLTVLEGDILDEPFLKRACQDVSVVIHTACIIDVFGVTHRESIMNVNVKGTQLLLEACVQASVPVFIYTSSIEVAGPNSYKEIIQNGHEEEPLENTWPTPYPYSKKLAEKAVLAANGWNLKNGDTLYTCALRPTYIYGEGGPFLSASINEALNNNGILSSVGKFSTVNPVYVGNVAWAHILALRALRDPKKAPSVRGQFYYISDDTPHQSYDNLNYILSKEFGLRLDSRWSLPL
Primary
Store at -20°C. Avoid freeze-thaw cycles.
Western Blot 1:100 - 1:500, Immunocytochemistry/ Immunofluorescence 1:10-1:100
Polyclonal
Purified
RUO
PBS with 50% glycerol, pH7.3.
3284
IgG
Affinity purified
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.