missing translation for 'onlineSavingsMsg'
Learn More

Beta-Hydroxy Thyroxine (>90%), TRC

Product Code. 30399712
Change view
Click to view available options
Quantity:
1 mg
10 mg
Unit Size:
1mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30399712 1 mg 1mg
30402619 10 mg -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30399712 Supplier Toronto Research Chemicals Supplier No. TRCH9636001MG
 Restricted Item
Estimated Shipment: 13-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 107849-54-7
Molecular Formula C15H11I4NO5
Molecular Weight (g/mol) 792.87
Synonym β-[3,5-Diiodo-4-(3,5-diiodo-4-hydroxyphenoxy)phenyl]serine, Levothyroxine beta-Hydroxy Impurity, Levothyroxine beta-Hydroxy T4 Impurity
SMILES IC1=C(OC2=CC(I)=C(O)C(I)=C2)C(I)=CC(C(O)C(N)C(O)=O)=C1

Specifications

Quantity 1 mg
InChI Formula InChI=1S/C15H11I4NO5/c16-7-3-6(4-8(17)13(7)22)25-14-9(18)1-5(2-10(14)19)12(21)11(20)15(23)24/h1-4,11-12,21-22H,20H2,(H,23,24)/t11-,12?/m0/s1
Chemical Name or Material beta-Hydroxy Thyroxine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.