CAS RN 107849-54-7
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Beta-Hydroxy Thyroxine (>90%), TRC
CAS: 107849-54-7 Molecular Formula: C15H11I4NO5 Molecular Weight (g/mol): 792.87 Synonym: β-[3,5-Diiodo-4-(3,5-diiodo-4-hydroxyphenoxy)phenyl]serine,Levothyroxine beta-Hydroxy Impurity,Levothyroxine beta-Hydroxy T4 Impurity SMILES: IC1=C(OC2=CC(I)=C(O)C(I)=C2)C(I)=CC(C(O)C(N)C(O)=O)=C1