missing translation for 'onlineSavingsMsg'
Learn More

Beta-D-Galactopyranosylamine, TRC

Product Code. 30395398
Change view
Click to view available options
Quantity:
100 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30395398 1 g
30390489 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30395398 Supplier Toronto Research Chemicals Supplier No. TRCG1544751G

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6318-23-6
Molecular Formula C6 H13 N O5
Molecular Weight (g/mol) 179.17
Synonym 1-Amino-1-Deoxy-beta-D-Galactose
IUPAC Name (2R,3R,4S,5R,6R)-2-amino-6-(hydroxymethyl)oxane-3,4,5-triol
SMILES N[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O

Specifications

Quantity 1 g
InChI Formula InChI=1S/C6H13NO5/c7-6-5(11)4(10)3(9)2(1-8)12-6/h2-6,8-11H,1,7H2/t2-,3+,4+,5-,6-/m1/s1
Formula Weight 179.0794
Chemical Name or Material beta-D-Galactopyranosylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.