CAS RN 6318-23-6
IUPAC Name:
—
Synonyms:
—
Molecular Weight (g/mol):
—
Molecular Formula:
—
InChi Key:
—
SMILES:
—
1
–
1
of
1
results
1
Beta-D-Galactopyranosylamine, TRC
CAS: 6318-23-6 Molecular Formula: C6 H13 N O5 Molecular Weight (g/mol): 179.17 Synonym: 1-Amino-1-Deoxy-beta-D-Galactose IUPAC Name: (2R,3R,4S,5R,6R)-2-amino-6-(hydroxymethyl)oxane-3,4,5-triol SMILES: N[C@@H]1O[C@H](CO)[C@H](O)[C@H](O)[C@H]1O