missing translation for 'onlineSavingsMsg'
Learn More

beta Casein Antibody (F20.14) - BSA Free, Novus Biologicals™

Mouse Monoclonal Antibody has been used in 2 publications

£169.00

Specifications

Antigen beta Casein
Clone F20.14
Concentration 0.88 mg/ml
Dilution Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500
Applications Western Blot
View More Specs

Products 1
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
18585161
View Documents
Novus Biologicals
NB100-2720-0.025mg
0.025 mg
£169.00
25µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

beta Casein Monoclonal specifically detects beta Casein in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications

Specifications

beta Casein
0.88 mg/ml
Western Blot
Unconjugated
Mouse
Human
1447
Purified casein and a-lactalbumin.
Primary
Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles.
28 kDa
F20.14
Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500
Monoclonal
Purified
RUO
beta-casein, CASB, casein beta
CSN2
IgG1
Protein A purified
Does not cross react with alpha-lactalbumin.
Videos
SDS
Documents

Documents

Product Certifications

For Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.