missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
beta Casein Monoclonal specifically detects beta Casein in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
Specifications
| Antigen | beta Casein |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | F20.14 |
| Concentration | 0.88 mg/ml |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1:100 - 1:2000, Immunohistochemistry 1:10 - 1:500 |
| Gene Alias | beta-casein, CASB, casein beta |
| Gene Symbols | CSN2 |
| Host Species | Mouse |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?