missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Arrestin 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£149.00
Specifications
| Antigen | beta-Arrestin 2 |
|---|---|
| Concentration | 1 mg/mL |
| Dilution | Western Blot 1:2000, Immunoprecipitation 1:10 - 1:500 |
| Applications | Western Blot |
| Classification | Polyclonal |
Description
beta-Arrestin 2 Polyclonal specifically detects beta-Arrestin 2 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunoprecipitation.Specifications
| beta-Arrestin 2 | |
| Western Blot 1:2000, Immunoprecipitation 1:10 - 1:500 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| P32121 | |
| 409 | |
| Synthetic peptide corresponding to residues C D(384) D I V F E D F A R L R L K(397) of human beta-arrestin 2. | |
| Primary | |
| Store at -20°C. Avoid freeze/thaw cycles. |
| 1 mg/mL | |
| Western Blot | |
| Unconjugated | |
| RUO | |
| PBS with 1 mg/ml BSA with 0.05% Sodium Azide | |
| ARB2, ARR2, arrestin 3, Arrestin beta-2, arrestin, beta 2, BARR2, beta-arrestin-2, DKFZp686L0365 | |
| ARRB2 | |
| IgG | |
| Affinity Purified | |
| Detects recombinant rat beta-arrestin and beta-arrestin 2. This does not detect visual or cone arrestin. |
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title