missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta-Arrestin 2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB300-587-0.025mg
This item is not returnable.
View return policy
Description
beta-Arrestin 2 Polyclonal specifically detects beta-Arrestin 2 in Human, Mouse, Rat, Bovine samples. It is validated for Western Blot, Immunoprecipitation.
Specifications
| beta-Arrestin 2 | |
| Polyclonal | |
| Unconjugated | |
| PBS with 1 mg/ml BSA with 0.05% Sodium Azide | |
| ARB2, ARR2, arrestin 3, Arrestin beta-2, arrestin, beta 2, BARR2, beta-arrestin-2, DKFZp686L0365 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 409 | |
| Human | |
| IgG |
| Western Blot | |
| 1 mg/mL | |
| Western Blot 1:2000, Immunoprecipitation 1:10 - 1:500 | |
| P32121 | |
| ARRB2 | |
| Synthetic peptide corresponding to residues C D(384) D I V F E D F A R L R L K(397) of human beta-arrestin 2. | |
| 0.025 mg | |
| Primary | |
| Detects recombinant rat beta-arrestin and beta-arrestin 2. This does not detect visual or cone arrestin. | |
| Store at -20°C. Avoid freeze/thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction