missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat beta-Actin Antibody, R&D Systems™

Mouse Monoclonal Antibody
£189.00 - £391.00
Specifications
| Antigen | beta-Actin |
|---|---|
| Clone | 937215 |
| Dilution | Western Blot 0.01 ug/mL, Simple Western 1 ug/mL, Immunocytochemistry 0.1-25 ug/mL |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
|---|---|---|---|---|---|---|---|---|---|
| Produktcode | Marke | Quantity | Preis | Menge & Verfügbarkeit | |||||
18794068
![]() |
R&D Systems
MAB8929-SP |
25 μg |
£189.00
25µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18781627
|
R&D Systems
MAB8929 |
100 μg |
£391.00
100µg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Beschreibung
beta-Actin Monoclonal specifically detects beta-Actin in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.Spezifikation
| beta-Actin | |
| Western Blot 0.01 ug/mL, Simple Western 1 ug/mL, Immunocytochemistry 0.1-25 ug/mL | |
| Monoclonal | |
| Purified | |
| RUO | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, betaActin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1 | |
| ACTB | |
| IgG1 | |
| Protein A or G purified from hybridoma culture supernatant | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| 937215 | |
| Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Mouse | |
| Human, Mouse, Rat | |
| P60709 | |
| 60 | |
| Peptide containing a sequence at the N-terminus of human beta-Actin Accession # P60709 | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Detects human, mouse, and rat beta-Actin in Western blots. |
For Research Use Only
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts