missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human/Mouse/Rat beta-Actin Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB8929
This item is not returnable.
View return policy
Description
beta-Actin Monoclonal specifically detects beta-Actin in Human, Mouse, Rat samples. It is validated for Western Blot, Simple Western, Immunocytochemistry.
Specifications
| beta-Actin | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ACTB, actin, beta, actin, cytoplasmic 1, B-Actin, Beta Actin, beta cytoskeletal actin, betaActin, Beta-actin, BRWS1, I(2)-Actin, PS1TP5-binding protein 1, PS1TP5BP1 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 60 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Western Blot, Immunocytochemistry | |
| 937215 | |
| Western Blot 0.01 ug/mL, Simple Western 1 ug/mL, Immunocytochemistry 0.1-25 ug/mL | |
| P60709 | |
| ACTB | |
| Peptide containing a sequence at the N-terminus of human beta-Actin Accession # P60709 | |
| 100 μg | |
| Primary | |
| Detects human, mouse, and rat beta-Actin in Western blots. | |
| Human, Mouse, Rat | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction