missing translation for 'onlineSavingsMsg'
Learn More
Learn More
beta 2-Microglobulin Antibody (246-E9.E7 (same as HLA.ABC.m2)) - Azide and BSA Free, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-47700-0.1MG
This item is not returnable.
View return policy
Description
beta 2-Microglobulin Monoclonal specifically detects beta 2-Microglobulin in Human, Primate (Negative) samples. It is validated for Flow Cytometry, Immunofluorescence, CyTOF-ready.
Specifications
| beta 2-Microglobulin | |
| Monoclonal | |
| 1.0 mg/mL | |
| Flow Cytometry : 0.5 - 1 ug/million cells in 0.1 ml, Immunofluorescence : 0.5 - 1.0 ug/ml, CyTOF-ready | |
| beta chain of MHC class I molecules, beta-2-microglobin, beta-2-microglobulin | |
| Mouse | |
| 12 kDa | |
| 0.1 mg | |
| Adaptive Immunity, Cancer, Cell Biology, Diabetes Research, Immunology | |
| 567 | |
| Human, Primate (Negative) | |
| Purified |
| Flow Cytometry, Immunofluorescence, CyTOF | |
| 246-E9.E7 (same as HLA.ABC.m2) | |
| Unconjugated | |
| PBS with No Preservative | |
| B2M | |
| Human PBL s from a T-cell acute lymphoblastic leukemia (T-ALL) patient. | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Recognizes a protein of 12kDa, identified as beta-microglobulin. beta-microglobulin non-covalently associates with the 44kDa chain to form the HLA Class I antigen complex. Human beta-2 microglobulin associated with HLA Class I antigens is expressed on many types of cells including lymphocytes, thymocytes, monocytes, granulocytes, platelets, endothelial cells, and epithelial cells. It is absent on erythrocytes. This MAb is specific to human beta-2 microglobulin and does not react with non-human primate cells. This antibody reacts with all cell types excluding erythrocytes. The detection of beta-2 microglobulin in body fluids has been used as a tumor marker and for monitoring patients with HIV infection. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction