missing translation for 'onlineSavingsMsg'
Learn More

Berberine chloride hydrate, 96%, water <17%, Thermo Scientific™

£20.40 - £54.20

Chemical Identifiers

CAS 141433-60-5
Molecular Formula C20H20ClNO5
Molecular Weight (g/mol) 389.832
MDL Number MFCD00149998
InChI Key BPNJXFPOPCFZOC-UHFFFAOYSA-M
Synonym berberine chloride hydrate, 9,10-dimethoxy-5,6-dihydro-1,3 dioxolo 4,5-g isoquinolino 3,2-a isoquinolin-7-ium chloride hydrate, c20h18no4.cl.h2o, benzo g-1,3-benzodioxolo 5,6-a quinolizinium, 5,6-dihydro-9,10-dimethoxy-, chloride, monohydrate, kyoberin tn, 9,10-dimethoxy-5,6-dihydro-2h-1,3-dioxolano 4,5-g isoquinolino 3,2-a isoquinol ine, chloride, hydrate, berberinechloridehydrate, berberine chloride xhydrate, berberine chloride hydrate jp17
PubChem CID 155074
SMILES COC1=C(C2=C[N+]3=C(C=C2C=C1)C4=CC5=C(C=C4CC3)OCO5)OC.O.[Cl-]
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
11396656
SDS View Documents
Thermo Scientific Alfa Aesar
L03807.06
5g
£20.40
5g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
11306666
SDS View Documents
Thermo Scientific Alfa Aesar
L03807.14
25g
£54.20
25g
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Primarily for the treatment of gastroenteritis, dysentery and other intestinal infections and conjunctivitis, purulent otitis media.

Solubility
Soluble in water.

Notes
Light sensitive. Store in the dark. Store away from oxidizing agents, light.
Specifications

Chemical Identifiers

141433-60-5
389.832
BPNJXFPOPCFZOC-UHFFFAOYSA-M
155074
C20H20ClNO5
MFCD00149998
berberine chloride hydrate, 9,10-dimethoxy-5,6-dihydro-1,3 dioxolo 4,5-g isoquinolino 3,2-a isoquinolin-7-ium chloride hydrate, c20h18no4.cl.h2o, benzo g-1,3-benzodioxolo 5,6-a quinolizinium, 5,6-dihydro-9,10-dimethoxy-, chloride, monohydrate, kyoberin tn, 9,10-dimethoxy-5,6-dihydro-2h-1,3-dioxolano 4,5-g isoquinolino 3,2-a isoquinol ine, chloride, hydrate, berberinechloridehydrate, berberine chloride xhydrate, berberine chloride hydrate jp17
COC1=C(C2=C[N+]3=C(C=C2C=C1)C4=CC5=C(C=C4CC3)OCO5)OC.O.[Cl-]

Specifications

141433-60-5
MFCD00149998
Light Sensitive
berberine chloride hydrate, 9,10-dimethoxy-5,6-dihydro-1,3 dioxolo 4,5-g isoquinolino 3,2-a isoquinolin-7-ium chloride hydrate, c20h18no4.cl.h2o, benzo g-1,3-benzodioxolo 5,6-a quinolizinium, 5,6-dihydro-9,10-dimethoxy-, chloride, monohydrate, kyoberin tn, 9,10-dimethoxy-5,6-dihydro-2h-1,3-dioxolano 4,5-g isoquinolino 3,2-a isoquinol ine, chloride, hydrate, berberinechloridehydrate, berberine chloride xhydrate, berberine chloride hydrate jp17
BPNJXFPOPCFZOC-UHFFFAOYSA-M
389.832
371.82 (Anhydrous)
water <17%
C20H20ClNO5
3836585
14,1154
Soluble in water.
COC1=C(C2=C[N+]3=C(C=C2C=C1)C4=CC5=C(C=C4CC3)OCO5)OC.O.[Cl-]
155074
97%
Berberine chloride hydrate
Videos
Safety and Handling

Safety and Handling

missing translation for 'einecsNumber' : 211-195-9

missing translation for 'rtecsNumber' : DR9866400

missing translation for 'tsca' : Yes

SDS
Documents

Documents

Product Certifications

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.