missing translation for 'onlineSavingsMsg'
Learn More

Berberine chloride hydrate, 96%, water <17%, Thermo Scientific™

Brand:  Thermo Scientific Alfa Aesar L03807.06

Quantity 5g

 View more versions of this product

Product Code. 11396656

  • £20.40 / 5g

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
Primarily for the treatment of gastroenteritis, dysentery and other intestinal infections and conjunctivitis, purulent otitis media.

Solubility
Soluble in water.

Notes
Light sensitive. Store in the dark. Store away from oxidizing agents, light.
TRUSTED_SUSTAINABILITY
Specifications

Chemical Identifiers

141433-60-5
389.832
BPNJXFPOPCFZOC-UHFFFAOYSA-M
155074
C20H20ClNO5
MFCD00149998
berberine chloride hydrate, 9,10-dimethoxy-5,6-dihydro-1,3 dioxolo 4,5-g isoquinolino 3,2-a isoquinolin-7-ium chloride hydrate, c20h18no4.cl.h2o, benzo g-1,3-benzodioxolo 5,6-a quinolizinium, 5,6-dihydro-9,10-dimethoxy-, chloride, monohydrate, kyoberin tn, 9,10-dimethoxy-5,6-dihydro-2h-1,3-dioxolano 4,5-g isoquinolino 3,2-a isoquinol ine, chloride, hydrate, berberinechloridehydrate, berberine chloride xhydrate, berberine chloride hydrate jp17
COC1=C(C2=C[N+]3=C(C=C2C=C1)C4=CC5=C(C=C4CC3)OCO5)OC.O.[Cl-]

Specifications

141433-60-5
MFCD00149998
3836585
14,1154
Soluble in water.
COC1=C(C2=C[N+]3=C(C=C2C=C1)C4=CC5=C(C=C4CC3)OCO5)OC.O.[Cl-]
155074
97%
Berberine chloride hydrate
C20H20ClNO5
5g
Light Sensitive
berberine chloride hydrate, 9,10-dimethoxy-5,6-dihydro-1,3 dioxolo 4,5-g isoquinolino 3,2-a isoquinolin-7-ium chloride hydrate, c20h18no4.cl.h2o, benzo g-1,3-benzodioxolo 5,6-a quinolizinium, 5,6-dihydro-9,10-dimethoxy-, chloride, monohydrate, kyoberin tn, 9,10-dimethoxy-5,6-dihydro-2h-1,3-dioxolano 4,5-g isoquinolino 3,2-a isoquinol ine, chloride, hydrate, berberinechloridehydrate, berberine chloride xhydrate, berberine chloride hydrate jp17
BPNJXFPOPCFZOC-UHFFFAOYSA-M
389.832
371.82 (Anhydrous)
water <17%
Product Suggestions

Product Suggestions

Videos
SDS Documents

Documents

Certificates
Promotions

Promotions

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.